Assay Details
Target Gene Details
Entrez Gene ID: | 432508 |
Gene Name: | cleavage and polyadenylation specific factor 6 |
Gene Aliases: |
4733401N12Rik, AI256641, CFIM, CFIM68, HPBRII-4, HPBRII-7 |
Location: |
Chr.10:117344667-117376986 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpsf6 | NM_001013391.2 | 2 | 280 | NP_001013409.1 |
| AK046856.1 | 2 | 236 | BAC32898.1 | |
| AK048615.1 | 2 | 214 | BAC33392.1 | |
| AK049749.1 | 2 | 278 | ||
| AK077684.1 | 2 | 281 | ||
| AK168764.1 | 2 | 245 | BAE40600.1 | |
| AK199581.1 | ||||
| AK199961.1 | ||||
| BC068133.1 | 2 | 256 | AAH68133.1 |
More Information
Set Membership: |
|

Genomic Map