Assay Details
Target Gene Details
Entrez Gene ID: | 69412 |
Gene Name: | RIKEN cDNA 1700016L04 gene |
Gene Aliases: |
- |
Location: |
Chr.10:14705609-14759019 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700016L04Rik | NR_045824.1 | |||
| AK006045.1 |
Target Gene Details
Entrez Gene ID: | 76743 |
Gene Name: | gap junction protein, epsilon 1 |
Gene Aliases: |
AEY12, Cx23, D230044M03Rik, Gjf1, Gsfaey12 |
Location: |
Chr.10:14715626-14718214 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gje1 | NM_029722.1 | NP_083998.1 | ||
| AK018698.1 | BAB31350.1 |
More Information
Set Membership: |
|

Genomic Map