Assay Details
Target Gene Details
Entrez Gene ID: | 103220 |
Gene Name: | tetratricopeptide repeat domain 41 |
Gene Aliases: |
AI449705, AU041509, Gnn |
Location: |
Chr.10:86705632-86776844 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc41 | NM_001003910.2 | NP_001003910.2 | ||
| AK133195.1 | ||||
| AK166553.1 | BAE38851.1 | |||
| AY651021.1 | AAT81233.1 |

Genomic Map