Assay Details
Target Gene Details
Entrez Gene ID: | 103220 |
Gene Name: | tetratricopeptide repeat domain 41 |
Gene Aliases: |
AI449705, AU041509, Gnn |
Location: |
Chr.10:86705632-86776844 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc41 | NM_001003910.2 | 1 | 62 | NP_001003910.2 |
| NM_001003939.2 | 1 | 62 | NP_001003939.2 | |
| AK133195.1 | ||||
| AY651019.1 | 1 | 61 | AAT81231.1 | |
| BC030307.1 | AAH30307.1 |

Genomic Map