Assay Details
Target Gene Details
Entrez Gene ID: | 56530 |
Gene Name: | canopy 2 homolog (zebrafish) |
Gene Aliases: |
5330432A10Rik, AW229003, D10Bwg1546e, Tmem4, Zsig9 |
Location: |
Chr.10:128322431-128327187 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnpy2 | NM_019953.1 | NP_064337.1 | ||
| AF186115.1 | AAF01433.1 | |||
| AK007914.1 | BAB25346.1 | |||
| AK013014.1 | BAB28597.1 | |||
| AK013568.1 | BAB28909.1 | |||
| AK019927.1 | BAB31921.1 | |||
| AK150953.1 | BAE29986.1 | |||
| AK179967.1 | ||||
| AK213625.1 | ||||
| BC008261.1 | AAH08261.1 |

Genomic Map