Assay Details
Target Gene Details
Entrez Gene ID: | 103220 |
Gene Name: | tetratricopeptide repeat domain 41 |
Gene Aliases: |
AI449705, AU041509, Gnn |
Location: |
Chr.10:86705632-86776844 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc41 | NM_001003910.2 | NP_001003910.2 | ||
| NM_001003939.2 | NP_001003939.2 | |||
| NM_153595.3 | NP_705823.3 | |||
| AK133195.1 | ||||
| AY651019.1 | AAT81231.1 | |||
| AY651020.1 | AAT81232.1 | |||
| AY651021.1 | AAT81233.1 | |||
| BC030307.1 | AAH30307.1 |

Genomic Map