Assay Details
Target Gene Details
Entrez Gene ID: | 216292 |
Gene Name: | methyltransferase like 25 |
Gene Aliases: |
- |
Location: |
Chr.10:105763185-105841380 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl25 | NM_207522.2 | NP_997405.2 | ||
| AK144620.1 | BAE25973.1 | |||
| AK148535.1 | ||||
| BC067068.1 | AAH67068.1 |
More Information
Set Membership: |
|

Genomic Map