Assay Details
Target Gene Details
Entrez Gene ID: | 12974 |
Gene Name: | citrate synthase |
Gene Aliases: |
2610511A05Rik, 9030605P22Rik, Ahl4, BB234005, Cis |
Location: |
Chr.10:128337832-128362479 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cs | NM_026444.3 | NP_080720.1 | ||
| AB056479.1 | BAB63945.1 | |||
| AK010298.1 | ||||
| AK012151.1 | BAB28063.1 | |||
| AK145643.1 | BAE26560.1 | |||
| AK149990.1 | BAE29218.1 | |||
| AK163273.1 | BAE37268.1 | |||
| AK166814.1 | BAE39041.1 | |||
| AK167125.1 | BAE39273.1 | |||
| BC013554.1 | AAH13554.1 | |||
| BC029754.1 | AAH29754.1 |
More Information
Set Membership: |
|

Genomic Map