Assay Details
Target Gene Details
Entrez Gene ID: | 71902 |
Gene Name: | cullin associated and neddylation disassociated 1 |
Gene Aliases: |
2310038O07Rik, 6330512O03Rik, AI195005, AI846556, D10Ertd516e, Tp120a, mKIAA0829 |
Location: |
Chr.10:119198812-119240055 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cand1 | NM_027994.1 | 12 | 3463 | NP_082270.1 |
| AK009683.1 | 1 | 102 | BAB26438.1 | |
| AK078117.1 | 3 | 690 | ||
| AK129225.1 | 12 | 3424 | BAC98035.1 | |
| BC057457.1 | 5 | 2106 | AAH57457.1 |
More Information
Set Membership: |
|

Genomic Map