Assay Details
Target Gene Details
Entrez Gene ID: | 72567 |
Gene Name: | BCL2-associated transcription factor 1 |
Gene Aliases: |
2610102K23Rik, 2700025J07Rik, 2810454G14Rik, 5730534O06Rik, AI450190, AW556225, Btf, mKIAA0164 |
Location: |
Chr.10:20311461-20342503 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bclaf1 | NM_001025392.1 | 12 | 2892 | NP_001020563.1 |
| NM_001025393.1 | 11 | 2745 | NP_001020564.1 | |
| NM_153787.2 | 12 | 2886 | NP_722482.1 | |
| AK079110.1 | 8 | 1142 | BAC37542.1 | |
| AK088741.1 | 12 | 2842 | BAC40542.1 | |
| AK129071.1 | 9 | 2176 | BAC97881.1 | |
| AK149724.1 | BAE29048.1 | |||
| AK150017.1 | BAE29242.1 | |||
| AK161692.1 | 12 | 2905 | BAE36535.1 | |
| AK169491.1 | 1 | 41 | BAE41198.1 | |
| BC034300.1 | 12 | 2818 | AAH34300.1 | |
| CN530237.1 | 3 | 277 |
More Information
Set Membership: |
|

Genomic Map