Assay Details
Target Gene Details
Entrez Gene ID: | 72567 |
Gene Name: | BCL2-associated transcription factor 1 |
Gene Aliases: |
2610102K23Rik, 2700025J07Rik, 2810454G14Rik, 5730534O06Rik, AI450190, AW556225, Btf, mKIAA0164 |
Location: |
Chr.10:20311461-20342503 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bclaf1 | NM_001025392.1 | NP_001020563.1 | ||
| NM_001025393.1 | NP_001020564.1 | |||
| NM_153787.2 | NP_722482.1 | |||
| AK011802.1 | BAB27851.1 | |||
| AK082427.1 | 1 | 2190 | ||
| AK088741.1 | BAC40542.1 | |||
| AK141834.1 | BAE24851.1 | |||
| AK149724.1 | BAE29048.1 | |||
| AK150017.1 | BAE29242.1 | |||
| AK161692.1 | BAE36535.1 | |||
| AK162896.1 | BAE37104.1 | |||
| BC028963.1 | ||||
| BC034300.1 | AAH34300.1 | |||
| BY071148.1 |
More Information
Set Membership: |
|

Genomic Map