Assay Details
Target Gene Details
Entrez Gene ID: | 52468 |
Gene Name: | CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2 |
Gene Aliases: |
AI586070, D10Ertd73e, OS-4, OS4, SCP2 |
Location: |
Chr.10:126978717-126999975 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctdsp2 | NM_001113470.1 | NP_001106941.1 | ||
| NM_146012.2 | NP_666124.1 | |||
| AK045068.1 | ||||
| AK049271.1 | BAC33649.1 | |||
| AK170287.1 | ||||
| BC025650.1 | AAH25650.1 | |||
| BC085142.1 | AAH85142.1 | |||
| BC089307.1 | AAH89307.1 | |||
| CB527995.1 |

Genomic Map