Assay Details
Target Gene Details
Entrez Gene ID: | 67972 |
Gene Name: | ATPase, Ca++ transporting, plasma membrane 1 |
Gene Aliases: |
2810442I22Rik, E130111D10Rik, Pmca1 |
Location: |
Chr.10:98914359-99026143 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp2b1 | NM_026482.2 | 16 | 2932 | NP_080758.1 |
| BC029045.1 | 10 | 1572 | AAH29045.1 | |
| BC049262.1 | 10 | 1572 | AAH49262.1 |
More Information
Set Membership: |
|

Genomic Map