Assay Details
Target Gene Details
Entrez Gene ID: | 12365 |
Gene Name: | caspase 14 |
Gene Aliases: |
MICE, mini-ICE |
Location: |
Chr.10:78711995-78718347 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Casp14 | NM_009809.5 | 1 | 36 | NP_033939.1 |
| AK081645.1 | BAC38280.1 | |||
| BC101959.1 | AAI01960.1 | |||
| BC101960.1 | AAI01961.1 | |||
| BC103555.1 | AAI03556.1 | |||
| BC103556.1 | AAI03557.1 | |||
| BY134001.1 | 1 | 40 |
More Information
Set Membership: |
|

Genomic Map