Assay Details
Target Gene Details
Entrez Gene ID: | 216033 |
Gene Name: | catenin (cadherin associated protein), alpha 3 |
Gene Aliases: |
4930429L08Rik, 4933408A16, Catna3, Vr22 |
Location: |
Chr.10:63430098-65005395 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctnna3 | NM_001164376.1 | NP_001157848.1 | ||
| NM_001164519.1 | NP_001157991.1 | |||
| NM_177612.3 | NP_808280.2 | |||
| AF344871.1 | AAQ14965.1 | |||
| AK030166.1 | BAC26817.1 | |||
| AK142745.1 | BAE25182.1 | |||
| BC112409.1 | ||||
| DQ380429.1 | ||||
| DQ380430.1 |
More Information
Set Membership: |
|

Genomic Map