Assay Details
Target Gene Details
Entrez Gene ID: | 28169 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 3 |
Gene Aliases: |
AW061257, AW493985, D10Jhu12e, lpaat3 |
Location: |
Chr.10:78269174-78352484 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agpat3 | NM_053014.3 | NP_443747.2 | ||
| AK008965.1 | ||||
| AK075715.1 | BAC35905.1 | |||
| AK196461.1 | ||||
| BC052382.1 | AAH52382.1 | |||
| BC058519.1 | AAH58519.1 |
More Information
Set Membership: |
|

Genomic Map