Assay Details
Target Gene Details
Entrez Gene ID: | 216119 |
Gene Name: | ybeY metallopeptidase |
Gene Aliases: |
1110068N23Rik, A130042E20Rik, C21orf57, ORF57, ORF66 |
Location: |
Chr.10:76459567-76469114 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ybey | NM_172550.4 | 2 | 417 | NP_766138.3 |
| AK037727.1 | 2 | 144 | BAC29859.1 | |
| AK051909.1 | 1 | 554 | ||
| AK136543.1 | 2 | 168 | BAE23037.1 | |
| BC057308.1 | 2 | 417 | AAH57308.1 |
More Information
Set Membership: |
|

Genomic Map