Assay Details
Target Gene Details
Entrez Gene ID: | 22295 |
Gene Name: | cadherin 23 (otocadherin) |
Gene Aliases: |
4930542A03Rik, USH1D, ahl, ahl1, bob, bus, mdfw, nmf112, nmf181, nmf252, sals, v |
Location: |
Chr.10:60302414-60696515 on Build GRCm38 |
Assay Gene Location: | Within Exon 69 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh23 | NM_001252635.1 | 68 | 9979 | NP_001239564.1 |
| NM_023370.3 | 69 | 9985 | NP_075859.2 | |
| NR_045556.1 | 68 | 9979 | ||
| AF308939.1 | 67 | 9578 | AAG52817.1 | |
| AJ888000.1 | 66 | 9572 | CAI59609.1 | |
| AJ888001.1 | 66 | 9572 | CAI59610.1 | |
| AJ888002.1 | 66 | 9572 | CAI59611.1 | |
| AK039126.1 | 24 | 3320 | ||
| AY026062.1 | 67 | 9587 | AAK07670.1 | |
| AY563159.1 | 2 | 660 | AAT72159.1 | |
| AY563160.1 | 2 | 660 | AAT72160.1 | |
| AY563163.1 | 21 | 3200 | AAT72163.1 | |
| AY563164.1 | 21 | 3200 | AAT72164.1 |
More Information
Set Membership: |
|

Genomic Map