Assay Details
Target Gene Details
Entrez Gene ID: | 103268 |
Gene Name: | centrosomal protein 57-like 1 |
Gene Aliases: |
2410017P07Rik, 4930484D11Rik, AI607327, cep57R |
Location: |
Chr.10:41718840-41809868 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep57l1 | NM_001243074.1 | 6 | 695 | NP_001230003.1 |
| NM_001243075.1 | 4 | 386 | NP_001230004.1 | |
| NM_026643.5 | 6 | 613 | NP_080919.3 | |
| NM_029132.2 | 6 | 681 | NP_083408.1 | |
| AF397908.1 | 6 | 609 | ||
| AK010530.1 | 6 | 632 | ||
| AK012710.1 | 4 | 332 | BAB28426.1 | |
| AK015619.1 | 5 | 596 | BAB29907.1 | |
| BC021375.1 | 6 | 705 | AAH21375.1 | |
| BC138230.1 | 6 | 616 | ||
| BQ553570.1 | ||||
| CD354087.1 | 3 | 363 |

Genomic Map