Assay Details
Target Gene Details
Entrez Gene ID: | 171580 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 1 |
Gene Aliases: |
MICAL, MICAL-1, Nical |
Location: |
Chr.10:41475860-41487030 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mical1 | NM_001164433.1 | NP_001157905.1 | ||
| NM_138315.2 | NP_612188.1 | |||
| AK154665.1 | ||||
| AK154930.1 | ||||
| AK157196.1 | ||||
| AK159369.1 | BAE35027.1 | |||
| AK170382.1 | ||||
| AK170827.1 | ||||
| AK170874.1 | ||||
| AK170881.1 | BAE42090.1 | |||
| AK171234.1 | BAE42330.1 | |||
| AK171411.1 | BAE42437.1 | |||
| AK171429.1 | BAE42447.1 | |||
| AK171479.1 | BAE42481.1 | |||
| AK171779.1 | ||||
| AK172562.1 | ||||
| AK172716.1 | ||||
| BC021477.1 | AAH21477.1 | |||
| BC034682.1 | AAH34682.1 |

Genomic Map