Assay Details
Target Gene Details
Entrez Gene ID: | 270685 |
Gene Name: | methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like |
Gene Aliases: |
2410004L15Rik, AI647056, Fthfsdc1 |
Location: |
Chr.10:3970441-4167081 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mthfd1l | NM_001170785.1 | NP_001164256.1 | ||
| NM_001170786.1 | NP_001164257.1 | |||
| NM_172308.4 | NP_758512.3 | |||
| AK028211.1 | BAE20438.1 | |||
| AK036284.1 | BAE20500.1 | |||
| AK038579.1 | BAC30053.1 | |||
| AK075931.1 | ||||
| AK160025.1 | BAE35568.1 |
More Information
Set Membership: |
|

Genomic Map