Assay Details
Target Gene Details
Entrez Gene ID: | 14708 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma 7 |
Gene Aliases: |
AI840417 |
Location: |
Chr.10:80948624-81014961 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gng7 | NM_001038655.1 | NP_001033744.1 | ||
| NM_010319.3 | NP_034449.2 | |||
| AK141541.1 | BAE24725.1 | |||
| AK147843.1 | BAE28176.1 | |||
| AK158963.1 | BAE34744.1 | |||
| AK177829.1 | ||||
| AY035844.1 | AAK61365.1 | |||
| BC034108.1 | AAH34108.1 | |||
| BY271138.1 |
More Information
Set Membership: |
|

Genomic Map