Assay Details
Target Gene Details
Entrez Gene ID: | 432486 |
Gene Name: | N-acetylglucosamine-1-phosphate transferase, alpha and beta subunits |
Gene Aliases: |
EG432486, mKIAA1208 |
Location: |
Chr.10:88379137-88447330 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnptab | NM_001004164.2 | NP_001004164.2 | ||
| AK140867.1 | BAE24503.1 | |||
| AK154710.1 | BAE32779.1 | |||
| AK173132.1 | BAD32410.1 | |||
| BC060638.1 | AAH60638.1 | |||
| L36434.1 | AAA65688.1 |

Genomic Map