Assay Details
Target Gene Details
Entrez Gene ID: | 67371 |
Gene Name: | general transcription factor IIIC, polypeptide 6, alpha |
Gene Aliases: |
2410016F19Rik, AU019813 |
Location: |
Chr.10:40247701-40258158 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf3c6 | NM_026113.4 | NP_080389.2 | ||
| AK005261.1 | BAB23913.1 | |||
| AK007816.1 | BAB25281.1 | |||
| AK010514.1 | ||||
| AK162053.1 | BAE36699.1 | |||
| AK168070.1 | BAE40045.1 | |||
| BC027260.1 | AAH27260.1 | |||
| BC048446.1 | AAH48446.1 |
More Information
Set Membership: |
|

Genomic Map