Assay Details
Target Gene Details
Entrez Gene ID: | 20503 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 7 |
Gene Aliases: |
4921534N07Rik, Mct2 |
Location: |
Chr.10:125219284-125389562 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc16a7 | DQ140166.1 |

Genomic Map