Assay Details
Target Gene Details
Entrez Gene ID: | 404634 |
Gene Name: | H2A histone family, member Y2 |
Gene Aliases: |
macroH2A2 |
Location: |
Chr.10:61738647-61784104 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2afy2 | NM_207000.2 | NP_996883.1 | ||
| AF336305.1 | AAK52472.1 | |||
| AK018355.1 | ||||
| AK020744.1 | BAB32199.1 | |||
| AK032636.1 | BAC27963.1 | |||
| AK078464.1 | BAC37288.1 | |||
| AK165020.1 | BAE38003.1 | |||
| BC045140.1 | AAH45140.1 | |||
| BC046794.1 | AAH46794.1 | |||
| BC107279.1 | AAI07280.1 | |||
| BC107280.1 | AAI07281.1 | |||
| BC127144.1 |
More Information
Set Membership: |
|

Genomic Map