Assay Details
Target Gene Details
Entrez Gene ID: | 54387 |
Gene Name: | minichromosome maintenance deficient 3 (S. cerevisiae) associated protein |
Gene Aliases: |
GANP, mKIAA0572 |
Location: |
Chr.10:76468971-76515859 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcm3ap | NM_019434.2 | 1 | 171 | NP_062307.2 |
| AJ006590.1 | 1 | 204 | CAB44241.1 | |
| AK051901.1 | 1 | 202 | BAC34807.1 | |
| AK122313.1 | 1 | 215 | BAC65595.1 | |
| BC052452.1 | 1 | 171 | AAH52452.1 |
Target Gene Details
Entrez Gene ID: | 216119 |
Gene Name: | ybeY metallopeptidase |
Gene Aliases: |
1110068N23Rik, A130042E20Rik, C21orf57, ORF57, ORF66 |
Location: |
Chr.10:76459567-76469114 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ybey | NM_172550.4 | NP_766138.3 | ||
| BC057308.1 | AAH57308.1 |
More Information
Set Membership: |
|

Genomic Map