Assay Details
Target Gene Details
Entrez Gene ID: | 54387 |
Gene Name: | minichromosome maintenance deficient 3 (S. cerevisiae) associated protein |
Gene Aliases: |
GANP, mKIAA0572 |
Location: |
Chr.10:76468971-76515859 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcm3ap | NM_019434.2 | 11 | 3038 | NP_062307.2 |
| AJ006590.1 | 11 | 3071 | CAB44241.1 | |
| AK052382.1 | 6 | 894 | BAC34967.1 | |
| AK122313.1 | 11 | 3082 | BAC65595.1 | |
| BC052452.1 | 11 | 3038 | AAH52452.1 | |
| BC067414.1 | 10 | 1973 | AAH67414.1 |
More Information
Set Membership: |
|

Genomic Map