Assay Details
Target Gene Details
Entrez Gene ID: | 17339 |
Gene Name: | major intrinsic protein of eye lens fiber |
Gene Aliases: |
Aqp0, Cat, Hfi, Lop, MIP26, Svl, shrivelled |
Location: |
Chr.10:128225838-128231811 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mip | NM_008600.4 | NP_032626.2 | ||
| AK053490.1 | BAC35401.1 | |||
| AK053494.1 | BAC35402.1 | |||
| AK136287.1 | BAE22916.1 | |||
| BC082567.1 | AAH82567.1 | |||
| U27502.1 | AAC52416.1 |
More Information
Set Membership: |
|

Genomic Map