Assay Details
Target Gene Details
Entrez Gene ID: | 75406 |
Gene Name: | NADH dehydrogenase (ubiquinone) Fe-S protein 7 |
Gene Aliases: |
1010001M04Rik |
Location: |
Chr.10:80249121-80256793 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufs7 | NM_029272.3 | NP_083548.1 | ||
| AK003132.1 | BAB22592.1 | |||
| BC013503.1 | AAH13503.1 |
More Information
Set Membership: |
|

Genomic Map