Assay Details
Target Gene Details
Entrez Gene ID: | 16414 |
Gene Name: | integrin beta 2 |
Gene Aliases: |
2E6, AI528527, Cd18, LAD, LCAMB, Lfa1, MF17 |
Location: |
Chr.10:77530328-77565681 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgb2 | NM_008404.4 | NP_032430.2 | ||
| AK088435.1 | BAC40350.1 | |||
| AK136502.1 | BAE23013.1 | |||
| AK161236.1 | ||||
| AK169587.1 | ||||
| BC145644.1 | ||||
| M31039.1 | AAA39325.1 | |||
| X14951.1 | CAA33077.1 |
More Information
Set Membership: |
|

Genomic Map