Assay Details
Target Gene Details
Entrez Gene ID: | 64451 |
Gene Name: | DIP2 disco-interacting protein 2 homolog A (Drosophila) |
Gene Aliases: |
4931420H10Rik, AI426328, Dip2, Kiaa0184-hp, mKIAA0184 |
Location: |
Chr.10:76263049-76345301 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 33 - Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dip2a | NM_001081419.2 | NP_001074888.2 | ||
| AB093213.2 | BAC41397.2 | |||
| AK019853.1 | ||||
| BC068227.1 | AAH68227.1 |

Genomic Map