Assay Details
Target Gene Details
Entrez Gene ID: | 208666 |
Gene Name: | DIRAS family, GTP-binding RAS-like 1 |
Gene Aliases: |
Di-Ras1, Gbts1, RIG |
Location: |
Chr.10:81019589-81025821 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Diras1 | NM_145217.2 | NP_660252.1 | ||
| AK081336.1 | ||||
| BC057556.1 | AAH57556.1 |
More Information
Set Membership: |
|

Genomic Map