Assay Details
Target Gene Details
Entrez Gene ID: | 67933 |
Gene Name: | host cell factor C2 |
Gene Aliases: |
1700129L13Rik, AU019838 |
Location: |
Chr.10:82697497-82795865 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hcfc2 |
Target Gene Details
Entrez Gene ID: | 18045 |
Gene Name: | nuclear transcription factor-Y beta |
Gene Aliases: |
AA985999, Cbf-A |
Location: |
Chr.10:82748700-82764148 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfyb | NM_010914.2 | NP_035044.1 | ||
| AK010762.1 | BAB27166.1 | |||
| AK011792.1 | BAB27844.1 | |||
| AK079209.1 | BAC37577.1 | |||
| AK146168.1 | BAE26948.1 | |||
| AK154749.1 | ||||
| AK155749.1 | BAE33416.1 | |||
| BB858171.1 | ||||
| BC010719.1 | AAH10719.1 | |||
| X55316.1 | CAA39024.1 |
More Information
Set Membership: |
|

Genomic Map