Assay Details
Target Gene Details
Entrez Gene ID: | 327762 |
Gene Name: | DNA replication helicase 2 homolog (yeast) |
Gene Aliases: |
Dna2l, E130315B21Rik |
Location: |
Chr.10:62946980-62974188 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dna2 | NM_177372.3 | 2 | 183 | NP_796346.2 |
| AK028381.1 | 2 | 160 | BAC25919.1 | |
| AK129051.1 | 2 | 168 | BAC97861.1 | |
| BC030382.1 | 2 | 125 | AAH30382.1 | |
| BC115716.1 | 2 | 137 | ||
| BY768981.1 | 2 | 185 |

Genomic Map