Assay Details
Target Gene Details
Entrez Gene ID: | 14609 |
Gene Name: | gap junction protein, alpha 1 |
Gene Aliases: |
AU042049, AW546267, Cnx43, Cx43, Cx43alpha1, Gja-1, Npm1, connexin43 |
Location: |
Chr.10:56377201-56390423 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gja1 | NM_010288.3 | 1 | 70 | NP_034418.1 |
| AK011459.1 | BAB27632.1 | |||
| AK036979.1 | BAC29656.1 | |||
| AK145692.1 | BAE26592.1 | |||
| AK161997.1 | ||||
| AK171955.1 | ||||
| AY427554.1 | AAR06891.1 | |||
| AY427555.1 | 1 | 97 | AAR06892.1 | |
| AY427560.1 | AAR06897.1 | |||
| BC006894.1 | AAH06894.1 | |||
| BC055375.1 | AAH55375.1 | |||
| BY061175.1 | 1 | 73 | ||
| M63801.1 | AAA53027.1 |
More Information
Set Membership: |
|

Genomic Map