Assay Details
Target Gene Details
Entrez Gene ID: | 56422 |
Gene Name: | Hbs1-like (S. cerevisiae) |
Gene Aliases: |
2810035F15Rik, AI326327, eRFS |
Location: |
Chr.10:21295979-21368889 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hbs1l | NM_001042593.1 | NP_001036058.1 | ||
| NM_001145209.1 | NP_001138681.1 | |||
| NM_019702.2 | NP_062676.2 | |||
| AK006626.1 | BAB24679.1 | |||
| AK012856.1 | BAB28515.1 | |||
| AK146678.1 | BAE27353.1 | |||
| AK166140.1 | BAE38593.1 | |||
| AK168671.1 | BAE40521.1 | |||
| AK169281.1 | BAE41039.1 | |||
| AK173091.1 | BAD32369.1 | |||
| BC010251.1 | AAH10251.1 | |||
| BC085235.1 | AAH85235.1 | |||
| BU510509.1 | ||||
| CJ082265.1 |
More Information
Set Membership: |
|

Genomic Map