Assay Details
Target Gene Details
Entrez Gene ID: | 56422 |
Gene Name: | Hbs1-like (S. cerevisiae) |
Gene Aliases: |
2810035F15Rik, AI326327, eRFS |
Location: |
Chr.10:21295979-21368889 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hbs1l | NM_001042593.1 | 9 | 1294 | NP_001036058.1 |
| NM_019702.2 | 9 | 1303 | NP_062676.2 | |
| AF087672.1 | 6 | 1164 | AAD23351.1 | |
| AK146678.1 | 9 | 1260 | BAE27353.1 | |
| AK166140.1 | 9 | 1257 | BAE38593.1 | |
| AK168671.1 | 9 | 1257 | BAE40521.1 | |
| AK169281.1 | 9 | 1257 | BAE41039.1 | |
| AK173091.1 | 9 | 1249 | BAD32369.1 | |
| BC010251.1 | 9 | 1228 | AAH10251.1 |
More Information
Set Membership: |
|

Genomic Map