Assay Details
Target Gene Details
Entrez Gene ID: | 13853 |
Gene Name: | epilepsy, progressive myoclonic epilepsy, type 2 gene alpha |
Gene Aliases: |
TG-B, Tg(TcraK,TcrbK)TG-BFlv |
Location: |
Chr.10:11342690-11457477 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epm2a | NM_010146.2 | 3 | 664 | NP_034276.2 |
| AF124044.1 | 3 | 704 | AAD26336.1 | |
| BY730431.1 | ||||
| CN532498.1 | 2 | 295 |

Genomic Map