Assay Details
Target Gene Details
Entrez Gene ID: | 74053 |
Gene Name: | glutamate receptor interacting protein 1 |
Gene Aliases: |
4931400F03Rik, GRIP, eb |
Location: |
Chr.10:119453835-120087267 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grip1 | NM_001277292.1 | NP_001264221.1 | ||
| NM_001277293.1 | NP_001264222.1 | |||
| NM_001277294.1 | NP_001264223.1 | |||
| NM_028736.2 | NP_083012.1 | |||
| NM_130891.2 | NP_570961.1 | |||
| NM_133442.2 | NP_597699.1 | |||
| AB051560.1 | BAB46929.1 | |||
| AB051561.1 | BAB46930.1 | |||
| AB051562.1 | BAB46931.1 | |||
| AK029845.1 | 10 | 1151 | ||
| AK039673.1 | ||||
| AK220553.1 | BAD90538.1 | |||
| BC067398.1 | AAH67398.1 | |||
| BC072632.1 | AAH72632.1 |

Genomic Map