Assay Details
Target Gene Details
Entrez Gene ID: | 319415 |
Gene Name: | heparan sulfate (glucosamine) 3-O-sulfotransferase 5 |
Gene Aliases: |
D930005L05Rik, Gm1151, Hs3ost5 |
Location: |
Chr.10:36506804-36834397 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hs3st5 | NM_001081208.2 | NP_001074677.1 | ||
| NM_001253355.1 | NP_001240284.1 | |||
| NM_001253356.1 | NP_001240285.1 | |||
| AK031910.1 | BAC27601.1 | |||
| BC147490.1 | ||||
| BC147493.1 | ||||
| BC150973.1 | ||||
| BC158004.1 |
More Information
Set Membership: |
|

Genomic Map