Assay Details
Target Gene Details
Entrez Gene ID: | 27400 |
Gene Name: | hydroxysteroid (17-beta) dehydrogenase 6 |
Gene Aliases: |
17betaHSD9, Hsd17b9, Rdh8 |
Location: |
Chr.10:127990933-128007636 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsd17b6 | NM_013786.2 | 5 | 884 | NP_038814.1 |
| AK008212.1 | 5 | 943 | BAB25536.1 | |
| BC021836.1 | 5 | 884 | AAH21836.1 |
More Information
Set Membership: |
|

Genomic Map