Assay Details
Target Gene Details
Entrez Gene ID: | 17311 |
Gene Name: | kit ligand |
Gene Aliases: |
Clo, Con, Gb, Kitlg, Mgf, SCF, SF, SLF, Sl, blz, contrasted |
Location: |
Chr.10:100015809-100100416 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kitl | NM_013598.2 | 4 | 473 | NP_038626.1 |
| AK018777.1 | 4 | 473 | BAB31402.1 | |
| AK041961.1 | 4 | 591 | BAC31113.1 | |
| AK134301.1 | 4 | 473 | BAE22091.1 | |
| AK165233.1 | 4 | 471 | BAE38091.1 | |
| BC011322.1 | 4 | 443 | AAH11322.1 | |
| M57647.1 | 4 | 425 | AAA39538.1 | |
| M59915.1 | 4 | 291 | AAA40095.1 | |
| M64262.1 | 4 | 345 | AAA39378.1 | |
| S40364.1 | 4 | 381 | AAB22554.2 | |
| S40534.1 | 4 | 381 | AAB22555.2 | |
| S40536.1 | 4 | 381 | AAB22556.2 | |
| U44725.1 | 4 | 473 | AAC52447.1 | |
| X68989.1 | 3 | 276 | CAA48778.1 | |
| X95379.1 | 4 | 311 | CAA64666.1 | |
| X95380.1 | 4 | 311 | ||
| X95381.1 | 4 | 311 | CAA64667.1 | |
| X99322.1 | 4 | 311 | CAA67698.1 | |
| Y10287.1 | 4 | 311 | CAA71329.1 |

Genomic Map