Assay Details
Target Gene Details
Entrez Gene ID: | 26396 |
Gene Name: | mitogen-activated protein kinase kinase 2 |
Gene Aliases: |
AA589381, MEK2, MK2, Prkmk2 |
Location: |
Chr.10:81105912-81124697 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map2k2 | NM_023138.4 | NP_075627.2 | ||
| AK009392.1 | BAB26261.1 | |||
| AK011247.1 | ||||
| AK080574.1 | BAC37945.1 | |||
| AK138130.1 | ||||
| AK143425.1 | BAE25372.1 | |||
| AK171584.1 | BAE42542.1 | |||
| AK172055.1 | BAE42803.1 | |||
| AK181608.1 | ||||
| AK186466.1 | ||||
| AK203356.1 | ||||
| BC014830.1 | AAH14830.1 | |||
| S68267.1 | AAC60678.1 |
More Information
Set Membership: |
|

Genomic Map