Assay Details
Target Gene Details
Entrez Gene ID: | 208266 |
Gene Name: | DOT1-like, histone H3 methyltransferase (S. cerevisiae) |
Gene Aliases: |
A630076O07, AW907654, Dot1, KMT4, mDot1 |
Location: |
Chr.10:80755129-80795461 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dot1l | NM_199322.1 | 5 | 700 | NP_955354.1 |
| AK035144.1 | 5 | 701 | ||
| AK220271.1 | 5 | 686 | BAD90196.1 | |
| AY196089.1 | 5 | 700 | AAP42293.1 | |
| AY376663.1 | 3 | 559 | AAQ84680.1 | |
| JF508521.1 | 5 | 462 |
More Information
Set Membership: |
|

Genomic Map