Assay Details
Target Gene Details
Entrez Gene ID: | 237339 |
Gene Name: | l(3)mbt-like 3 (Drosophila) |
Gene Aliases: |
AI481284, MBT-1 |
Location: |
Chr.10:26274470-26375989 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| L3mbtl3 | NM_172787.2 | NP_766375.1 | ||
| AK030412.1 | BAC26951.1 | |||
| AK045667.1 | BAC32449.1 | |||
| AK049597.1 | ||||
| AK129443.1 | BAC98253.1 | |||
| BC082309.1 | AAH82309.1 | |||
| BC085192.1 |
More Information
Set Membership: |
|

Genomic Map