Assay Details
Target Gene Details
Entrez Gene ID: | 320398 |
Gene Name: | leucine-rich repeats and immunoglobulin-like domains 3 |
Gene Aliases: |
9030421L11Rik, 9130004I02Rik, 9430095K15Rik, mKIAA3016 |
Location: |
Chr.10:125966219-126015359 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrig3 | NM_177152.5 | NP_796126.4 | ||
| AK018522.1 | ||||
| AK129481.1 | BAC98291.1 | |||
| AK137007.1 | ||||
| AY505341.1 | AAR98630.1 | |||
| BC065142.1 | AAH65142.1 |
More Information
Set Membership: |
|

Genomic Map