Assay Details
Target Gene Details
Entrez Gene ID: | 26408 |
Gene Name: | mitogen-activated protein kinase kinase kinase 5 |
Gene Aliases: |
7420452D20Rik, ASK, ASK1, MAPKKK5, Mekk5 |
Location: |
Chr.10:19934513-20142750 on Build GRCm38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map3k5 | NM_008580.4 | NP_032606.4 | ||
| AB006787.3 | BAA23648.3 | |||
| BC116627.1 | ||||
| BC116628.1 | ||||
| BC133697.1 |
More Information
Set Membership: |
|

Genomic Map