Assay Details
Target Gene Details
Entrez Gene ID: | 327744 |
Gene Name: | RIKEN cDNA E130307A14 gene |
Gene Aliases: |
- |
Location: |
Chr.10:39621411-39732007 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E130307A14Rik | NR_038037.1 | 1 | 100 | |
| AK049799.1 | 1 | 60 | ||
| AK087502.1 | ||||
| AK164126.1 | 1 | 60 |

Genomic Map