Assay Details
Target Gene Details
Entrez Gene ID: | 69736 |
Gene Name: | nucleoporin 37 |
Gene Aliases: |
2410003L22Rik, 2810039M17Rik |
Location: |
Chr.10:88146976-88178395 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup37 | NM_027191.2 | NP_081467.2 | ||
| NM_028334.4 | NP_082610.1 | |||
| AK010370.1 | BAB26888.1 | |||
| AK012890.1 | BAB28538.1 | |||
| AK040463.1 | BAC30597.1 | |||
| AK043277.1 | 1 | 86 | ||
| AK165537.1 | BAE38243.1 | |||
| BC011102.1 | AAH11102.1 | |||
| CJ072834.1 |
More Information
Set Membership: |
|

Genomic Map